Learn More
Abnova™ Human DRAP1 Full-length ORF (NP_006433.2, 1 a.a. - 205 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010589-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Transcriptional repression is a general mechanism for regulating transcriptional initiation in organisms ranging from yeast to humans. Accurate initiation of transcription from eukaryotic protein-encoding genes requires the assembly of a large multiprotein complex consisting of RNA polymerase II and general transcription factors such as TFIIA, TFIIB, and TFIID. DR1 is a repressor that interacts with the TATA-binding protein (TBP) of TFIID and prevents the formation of an active transcription complex by precluding the entry of TFIIA and/or TFIIB into the preinitiation complex. The protein encoded by this gene is a corepressor of transcription that interacts with DR1 to enhance DR1-mediated repression. The interaction between this corepressor and DR1 is required for corepressor function and appears to stabilize the TBP-DR1-DNA complex. [provided by RefSeq]
Sequence: MPSKKKKYNARFPPARIKKIMQTDEEIGKVAAAVPVIISRALELFLESLLKKACQVTQSRNAKTMTTSHLKQCIELEQQFDFLKDLVASVPDMQGDGEDNHMDGDKGARRGRKPGSGGRKNGGMGTKSKDKKLSGTDSEQEDESEDTDTDGEEETSQPPPQASHPSAHFQSPPTPFLPFASTLPLPPAPPGPSAPDEEDEEDYDSSpecifications
NP_006433.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
48.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NC2-alpha | |
DRAP1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
10589 | |
DRAP1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPSKKKKYNARFPPARIKKIMQTDEEIGKVAAAVPVIISRALELFLESLLKKACQVTQSRNAKTMTTSHLKQCIELEQQFDFLKDLVASVPDMQGDGEDNHMDGDKGARRGRKPGSGGRKNGGMGTKSKDKKLSGTDSEQEDESEDTDTDGEEETSQPPPQASHPSAHFQSPPTPFLPFASTLPLPPAPPGPSAPDEEDEEDYDS | |
RUO | |
DRAP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |