missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DR1 Partial ORF (NP_001929, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00001810-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16194171

  • 528.00€ / 25µg

Estimated Shipment: 08-11-2023
to see stock.
Add to basket

Description

Description

This gene encodes a TBP- (TATA box-binding protein) associated phosphoprotein that represses both basal and activated levels of transcription. The encoded protein is phosphorylated in vivo and this phosphorylation affects its interaction with TBP. This protein contains a histone fold motif at the amino terminus, a TBP-binding domain, and a glutamine- and alanine-rich region. The binding of DR1 repressor complexes to TBP-promoter complexes may establish a mechanism in which an altered DNA conformation, together with the formation of higher order complexes, inhibits the assembly of the preinitiation complex and controls the rate of RNA polymerase II transcription. [provided by RefSeq]

Sequence: MASSSGNDDDLTIPRAAINKMIKETLPNVRVANDARELVVNCCTEFIHLISSEANEICNKSEKKTISPEHVIQALESLGFGSYISEVKEVLQECKTVALK
Specifications

Specifications

NP_001929
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MASSSGNDDDLTIPRAAINKMIKETLPNVRVANDARELVVNCCTEFIHLISSEANEICNKSEKKTISPEHVIQALESLGFGSYISEVKEVLQECKTVALK
RUO
DR1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
1810
DR1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NC2/NC2-BETA
DR1
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers