missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DOCK1 Partial ORF (NP_001371, 698 a.a. - 803 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00001793-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16194141

  • 528.00€ / 25µg

Estimated Shipment: 10-11-2023
to see stock.
Add to basket

Description

Description

This gene product binds to the SH3 domain of CRK protein. It may regulate cell surface extension and may have a role in the cell surface extension of an engulfing cell around a dying cell during apoptosis. [provided by RefSeq]

Sequence: LETYIKKHFSATLAYTKLTKVLKNYVDGAEKPGVNEQLYKAMKALESIFKFIVRSRILFNQLYENKGEADFVESLLQLFRSINDMMSSMSDQTVRVKGAALKYLPT
Specifications

Specifications

NP_001371
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.4kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LETYIKKHFSATLAYTKLTKVLKNYVDGAEKPGVNEQLYKAMKALESIFKFIVRSRILFNQLYENKGEADFVESLLQLFRSINDMMSSMSDQTVRVKGAALKYLPT
RUO
DOCK1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
1793
DOCK1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DOCK180/ced5
DOCK1
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers