missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DNASE2B Full-length ORF (NP_490649.1, 1 a.a. - 153 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00058511-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16193103

  • 349.00€ / 10µg

Estimated Shipment: 07-11-2023
to see stock.
Add to basket

Description

Description

The protein encoded by this gene shares considerable sequence similarity to, and is structurally related to DNase II. The latter is a well characterized endonuclease that catalyzes DNA hydrolysis in the absence of divalent cations at acidic pH. Unlike DNase II which is ubiquitously expressed, expression of this gene product is restricted to the salivary gland and lungs. The gene has been localized to chromosome 1p22.3 adjacent (and in opposite orientation) to the uricase pseudogene. Two transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq]

Sequence: MPQLCTRASSSEIPGRLLTTLQSAQGQKFLHFAKSDSFLDDIFAAWMAQRLKTHLLTETWQRKRQELPSNCSLPYHVYNIKAIKLSRHSYFSSYQDHAKWCISQKGTKNRWTCIGDLNRSPHQAFRSGGFICTQNWQIYQAFQGLVLYYESCK
Specifications

Specifications

NP_490649.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
44.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DLAD
DNASE2B
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
58511
DNASE2B (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MPQLCTRASSSEIPGRLLTTLQSAQGQKFLHFAKSDSFLDDIFAAWMAQRLKTHLLTETWQRKRQELPSNCSLPYHVYNIKAIKLSRHSYFSSYQDHAKWCISQKGTKNRWTCIGDLNRSPHQAFRSGGFICTQNWQIYQAFQGLVLYYESCK
RUO
DNASE2B
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers