Learn More
Abnova™ Human DNAJB13 Partial ORF (NP_705842, 71 a.a. - 180 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00374407-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
DNAJA13 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins. For background information on the DNAJ family, see 608375.[supplied by OMIM]
Sequence: GLKGGIPLEFGSQTPWTTGYVFHGKPEKVFHEFFGGNNPFSEFFDAEGSEVDLNFGGLQGRGVKKQDPQVERDLYLSLEDLFFGCTKKIKISRRVLNEDGYSSTIKDKILSpecifications
NP_705842 | |
Liquid | |
374407 | |
DNAJB13 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ46748/TSARG5/TSARG6 | |
DNAJB13 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GLKGGIPLEFGSQTPWTTGYVFHGKPEKVFHEFFGGNNPFSEFFDAEGSEVDLNFGGLQGRGVKKQDPQVERDLYLSLEDLFFGCTKKIKISRRVLNEDGYSSTIKDKIL | |
RUO | |
DNAJB13 | |
Wheat Germ (in vitro) | |
GST |