Learn More
Abnova™ Human DMGDH Partial ORF (NP_037523.2, 556 a.a. - 665 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00029958-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes an enzyme involved in the catabolism of choline, catalyzing the oxidative demethylation of dimethylglycine to form sarcosine. The enzyme is found as a monomer in the mitochondrial matrix, and uses flavin adenine dinucleotide and folate as cofactors. Mutation in this gene causes dimethylglycine dehydrogenase deficiency, characterized by a fishlike body odor, chronic muscle fatigue, and elevated levels of the muscle form of creatine kinase in serum. [provided by RefSeq]
Sequence: ANVIPKVGFTNISHMLTPKGRVYAELTVSHQSPGEFLLITGSGSELHDLRWIEEEAVKGGYDVEIKNITDELGVLGVAGPQARKVLQKLTSEDLSDDVFKFLQTKSLKVSSpecifications
NP_037523.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ANVIPKVGFTNISHMLTPKGRVYAELTVSHQSPGEFLLITGSGSELHDLRWIEEEAVKGGYDVEIKNITDELGVLGVAGPQARKVLQKLTSEDLSDDVFKFLQTKSLKVS | |
RUO | |
DMGDH | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
29958 | |
DMGDH (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DMGDHD/ME2GLYDH | |
DMGDH | |
Recombinant | |
wheat germ expression system |