Learn More
Abnova™ Human DIP2A Partial ORF (NP_055966, 1 a.a. - 99 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00023181-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene may be involved in axon patterning in the central nervous system. This gene is not highly expressed. Several transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MADRGCPLEAAPLPAEVRESLAELELELSEGDITQKGYEKKRAKLLARYIPLIQGIDPSLQAENRIPGPSQTTAAAPKQQKSRPTASRDERFRSDVHTESpecifications
NP_055966 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MADRGCPLEAAPLPAEVRESLAELELELSEGDITQKGYEKKRAKLLARYIPLIQGIDPSLQAENRIPGPSQTTAAAPKQQKSRPTASRDERFRSDVHTE | |
RUO | |
DIP2A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
23181 | |
DIP2A (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C21orf106/DIP2 | |
DIP2A | |
Recombinant | |
wheat germ expression system |