Learn More
Abcam Human DICER1 knockout HEK293T cell line [4-25]
Product Code. 32143468
Change view
Click to view available options
Quantity:
1,000,000 Cells
2 x 1,000,000 Cells
Unit Size:
Each
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32143468 2 x 1,000,000 Cells Pack of 2
32141110 1,000,000 Cells Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 32143468 Supplier Abcam Supplier No. ab2911172x1000000Cells_
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
DICER1 KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Expression plasmids encoding a pair of TALEN proteins designed to cleave the human dicer (hdcr) gene at nucleotide (nt) 1095 of the hDcr cDNA sequence, in coding exon 11. Clone 4-25. ...
DICER1 KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Expression plasmids encoding a pair of TALEN proteins designed to cleave the human dicer (hdcr) gene at nucleotide (nt) 1095 of the hDcr cDNA sequence, in coding exon 11. Clone 4-25. Knockout. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 2 x 1,000,000 Cells