Learn More
Abnova™ Human DHX38 Partial ORF (NP_054722.2, 342 a.a. - 450 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009785-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. The protein encoded by this gene is a member of the DEAD/H box family of splicing factors. This protein resembles yeast Prp16 more closely than other DEAD/H family members. It is an ATPase and essential for the catalytic step II in pre-mRNA splicing process. [provided by RefSeq]
Sequence: YSSEDYVRRREQHLHKQKQKRISAQRRQINEDNERWETNRMLTSGVVHRLEVDEDFEEDNAAKVHLMVHNLVPPFLDGRIVFTKQPEPVIPVKDATSDLAIIARKGSQTSpecifications
NP_054722.2 | |
Liquid | |
9785 | |
DHX38 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DDX38/KIAA0224/PRP16/PRPF16 | |
DHX38 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
YSSEDYVRRREQHLHKQKQKRISAQRRQINEDNERWETNRMLTSGVVHRLEVDEDFEEDNAAKVHLMVHNLVPPFLDGRIVFTKQPEPVIPVKDATSDLAIIARKGSQT | |
RUO | |
DHX38 | |
Wheat Germ (in vitro) | |
GST |