Learn More
Abnova™ Human DHRS7 Full-length ORF (NP_057113.1, 1 a.a. - 339 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00051635-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Short-chain dehydrogenases/reductases (SDRs), such as DHRS7, catalyze the oxidation/reduction of a wide range of substrates, including retinoids and steroids (Haeseleer and Palczewski, 2000 [PubMed 10800688]).[supplied by OMIM]
Sequence: MNWELLLWLLVLCALLLLLVQLLRFLRADGDLTLLWAEWQGRRPEWELTDMVVWVTGASSGIGEELAYQLSKLGVSLVLSARRVHELERVKRRCLENGNLKEKDILVLPLDLTDTGSHEAATKAVLQEFGRIDILVNNGGMSQRSLCMDTSLDVYRKLIELNYLGTVSLTKCVLPHMIERKQGKIVTVNSILGIISVPLSIGYCASKHALRGFFNGLRTELATYPGIIVSNICPGPVQSNIVENSLAGEVTKTIGNNGDQSHKMTTSRCVRLMLISMANDLKEVWISEQPFLLVTYLWQYMPTWAWWITNKMGKKRIENFKSGVDADSSYFKIFKTKHDSpecifications
NP_057113.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
64.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CGI-86/SDR34C1/retDSR4/retSDR4 | |
DHRS7 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
51635 | |
DHRS7 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MNWELLLWLLVLCALLLLLVQLLRFLRADGDLTLLWAEWQGRRPEWELTDMVVWVTGASSGIGEELAYQLSKLGVSLVLSARRVHELERVKRRCLENGNLKEKDILVLPLDLTDTGSHEAATKAVLQEFGRIDILVNNGGMSQRSLCMDTSLDVYRKLIELNYLGTVSLTKCVLPHMIERKQGKIVTVNSILGIISVPLSIGYCASKHALRGFFNGLRTELATYPGIIVSNICPGPVQSNIVENSLAGEVTKTIGNNGDQSHKMTTSRCVRLMLISMANDLKEVWISEQPFLLVTYLWQYMPTWAWWITNKMGKKRIENFKSGVDADSSYFKIFKTKHD | |
RUO | |
DHRS7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |