Learn More
Abnova™ Human DHRS1 Full-length ORF (NP_612461.1, 1 a.a. - 313 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00115817-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the short-chain dehydrogenases/reductases (SDR) family. The encoded enzyme contains a conserved catalytic domain and likely functions as an oxidoreductase. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq]
Sequence: MAAPMNGQVCVVTGASRGIGRGIALQLCKAGATVYITGRHLDTLRVVAQEAQSLGGQCVPVVCDSSQESEVRSLFEQVDREQQGRLDVLVNNAYAGVQTILNTRNKAFWETPASMWDDINNVGLRGHYFCSVYGARLMVPAGQGLIVVISSPGSLQYMFNVPYGVGKAACDKLAADCAHELRRHGVSCVSLWPGIVQTELLKEHMAKEEVLQDPVLKQFKSAFSSAETTELSGKCVVALATDPNILSLSGKVLPSCDLARRYGLRDVDGRPVQDYLSLSSVLSHVSGLGWLASYLPSFLRVPKWIIALYTSKFSpecifications
NP_612461.1 | |
Liquid | |
115817 | |
DHRS1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAAPMNGQVCVVTGASRGIGRGIALQLCKAGATVYITGRHLDTLRVVAQEAQSLGGQCVPVVCDSSQESEVRSLFEQVDREQQGRLDVLVNNAYAGVQTILNTRNKAFWETPASMWDDINNVGLRGHYFCSVYGARLMVPAGQGLIVVISSPGSLQYMFNVPYGVGKAACDKLAADCAHELRRHGVSCVSLWPGIVQTELLKEHMAKEEVLQDPVLKQFKSAFSSAETTELSGKCVVALATDPNILSLSGKVLPSCDLARRYGLRDVDGRPVQDYLSLSSVLSHVSGLGWLASYLPSFLRVPKWIIALYTSKF | |
RUO | |
DHRS1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
60.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp586I0523/FLJ14250/FLJ25430/MGC20204/SDR19C1 | |
DHRS1 | |
Yes | |
wheat germ expression system |