missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DHDH Partial ORF (NP_055290, 235 a.a. - 334 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00027294-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16195836

  • 349.00€ / 10µg

Estimated Shipment: 07-11-2023
to see stock.
Add to basket

Description

Description

This gene encodes an enzyme that belongs to the family of dihydrodiol dehydrogenases, which exist in multiple forms in mammalian tissues and are involved in the metabolism of xenobiotics and sugars. These enzymes catalyze the NADP1-linked oxidation of transdihydrodiols of aromatic hydrocarbons to corresponding catechols. This enzyme is a dimeric dihydrodiol dehydrogenase, and it differs from monomeric dihydrodiol dehydrogenases in its high substrate specificity for trans-dihydrodiols of aromatic hydrocarbons in the oxidative direction. [provided by RefSeq]

Sequence: SNTASVSGTKGMAQLLNPCWCPTELVVKGEHKEFPLPPVPKDCNFDNGAGMSYEAKHVWECLRKGMKESPVIPLSESELLADILEEVRKAIGVTFPQDKR
Specifications

Specifications

NP_055290
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SNTASVSGTKGMAQLLNPCWCPTELVVKGEHKEFPLPPVPKDCNFDNGAGMSYEAKHVWECLRKGMKESPVIPLSESELLADILEEVRKAIGVTFPQDKR
RUO
DHDH
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
27294
DHDH (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HUM2DD
DHDH
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers