Learn More
Abnova™ Human DGCR2 Full-length ORF (NP_005128.1, 1 a.a. - 550 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009993-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Deletions of the 22q11.2 have been associated with a wide range of developmental defects (notably DiGeorge syndrome, velocardiofacial syndrome, conotruncal anomaly face syndrome and isolated conotruncal cardiac defects) classified under the acronym CATCH 22. The DGCR2 gene encodes a novel putative adhesion receptor protein, which could play a role in neural crest cells migration, a process which has been proposed to be altered in DiGeorge syndrome. [provided by RefSeq]
Sequence: MVPKADSGAFLLLFLLVLTVTEPLRPELRCNPGQFACRSGTIQCIPLPWQCDGWATCEDESDEANCPEVTGEVRPHHGKEAVDPRQGRARGGDPSHFHAVNVAQPVRFSSFLGKCPTGWHHYEGTASCYRVYLSGENYWDAAQTCQRLNGSLATFSTDQELRFVLAQEWDQPERSFGWKDQRKLWVGYQYVITGRNRSLEGRWEVAFKGSSEVFLPPDPIFASAMSENDNVFCAQLQCFHFPTLRHHDLHSWHAESCYEKSSFLCKRSQTCVDIKDNVVDEGFYFTPKGDDPCLSCTCHGGEPEMCVAALCERPQGCQQYRKDPKECCKFMCLDPDGNSLFDSMASGMRLVVSCISSFLILSLLLFMVHRLRQRRRERIESLIGANLHHFNLGRRIPGFDYGPDGFGTGLTPLHLSDDGEGGTFHFHDPPPPYTAYKYPDIGQPDDPPPPYEASIHPDSVFYDPADDDAFEPVEVSLPAPGDGGSEGALLRRLEQPLPTAGASLADLEDSADSSSALLVPPDPAQSGSTPAAEALPGGGRHSRSSLNTVVSpecifications
NP_005128.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
87.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DGS-C/DKFZp686I1730/IDD/KIAA0163/LAN/SEZ-12 | |
DGCR2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
9993 | |
DGCR2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MVPKADSGAFLLLFLLVLTVTEPLRPELRCNPGQFACRSGTIQCIPLPWQCDGWATCEDESDEANCPEVTGEVRPHHGKEAVDPRQGRARGGDPSHFHAVNVAQPVRFSSFLGKCPTGWHHYEGTASCYRVYLSGENYWDAAQTCQRLNGSLATFSTDQELRFVLAQEWDQPERSFGWKDQRKLWVGYQYVITGRNRSLEGRWEVAFKGSSEVFLPPDPIFASAMSENDNVFCAQLQCFHFPTLRHHDLHSWHAESCYEKSSFLCKRSQTCVDIKDNVVDEGFYFTPKGDDPCLSCTCHGGEPEMCVAALCERPQGCQQYRKDPKECCKFMCLDPDGNSLFDSMASGMRLVVSCISSFLILSLLLFMVHRLRQRRRERIESLIGANLHHFNLGRRIPGFDYGPDGFGTGLTPLHLSDDGEGGTFHFHDPPPPYTAYKYPDIGQPDDPPPPYEASIHPDSVFYDPADDDAFEPVEVSLPAPGDGGSEGALLRRLEQPLPTAGASLADLEDSADSSSALLVPPDPAQSGSTPAAEALPGGGRHSRSSLNTVV | |
RUO | |
DGCR2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |