Learn More
Abnova™ Human DGCR14 Full-length ORF (AAH37829.1, 1 a.a. - 395 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008220-P01.25ug
Description
This gene is located within the minimal DGS critical region (MDGCR) thought to contain the gene(s) responsible for a group of developmental disorders. These disorders include DiGeorge syndrome, velocardiofacial syndrome, conotruncal anomaly face syndrome, and some familial or sporadic conotruncal cardiac defects which have been associated with microdeletion of 22q11.2. The encoded protein may be a component of C complex spliceosomes, and the orthologous protein in the mouse localizes to the nucleus. [provided by RefSeq]
Sequence: MLGACFPNLSSGDVMVFGPGGLGQPVLHPVLVLGHHQQPVLGFGSKLVVGPVVGPQACLGVQFALSAVLALPLPLEGGRRRGFGLGGLQPRVAEDLIDVEPLADVGLQHAVDEVLALAGQVLGAREVHTVLLLDTQHLPDVGVVIGHGAADHDVQDHAQAPDVIHLGLVGDALQHLGGCICCRPAEGLAEDDAPIAVPQAALGEAKVRQLDVEVLVEEKVLALEVPVDDVQVVAVLDGGGELSEHLACHVLMQGSLALDELEKRLAPLPSTSPRHVGQQALSSFTLPEASGWAALGWASTQGWDTQHCLRVTSANELIKPRSGGGDWAEGEQGLWNRGSQAGLAGAPTPRPLEGSLSPGQDSGGPAAAALVSLSQVKLEGRRPTSSLFASPGCGDSpecifications
AAH37829.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
67kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 μg | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DGCR13/DGS-H/DGS-I/DGSH/DGSI/ES2/Es2el | |
DGCR14 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
8220 | |
DGCR14 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLGACFPNLSSGDVMVFGPGGLGQPVLHPVLVLGHHQQPVLGFGSKLVVGPVVGPQACLGVQFALSAVLALPLPLEGGRRRGFGLGGLQPRVAEDLIDVEPLADVGLQHAVDEVLALAGQVLGAREVHTVLLLDTQHLPDVGVVIGHGAADHDVQDHAQAPDVIHLGLVGDALQHLGGCICCRPAEGLAEDDAPIAVPQAALGEAKVRQLDVEVLVEEKVLALEVPVDDVQVVAVLDGGGELSEHLACHVLMQGSLALDELEKRLAPLPSTSPRHVGQQALSSFTLPEASGWAALGWASTQGWDTQHCLRVTSANELIKPRSGGGDWAEGEQGLWNRGSQAGLAGAPTPRPLEGSLSPGQDSGGPAAAALVSLSQVKLEGRRPTSSLFASPGCGD | |
RUO | |
DGCR14 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.