missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DGAT2 Partial ORF (NP_115953.2, 289 a.a. - 388 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00084649-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16100817

  • 508.00€ / 25µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

Acyl-CoA:diacylglycerol acyltransferase, or DGAT (EC 2.3.1.20), is responsible for the synthesis of triglycerides. It catalyzes a reaction in which diacylglycerol is covalently joined to long chain fatty acyl-CoAs.[supplied by OMIM]

Sequence: IFEEGSWGRWVQKKFQKYIGFAPCIFHGRGLFSSDTWGLVPYSKPITTVVGEPITIPKLEHPTQQDIDLYHTMYMEALVKLFDKHKTKFGLPETEVLEVN
Specifications

Specifications

NP_115953.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
IFEEGSWGRWVQKKFQKYIGFAPCIFHGRGLFSSDTWGLVPYSKPITTVVGEPITIPKLEHPTQQDIDLYHTMYMEALVKLFDKHKTKFGLPETEVLEVN
RUO
DGAT2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
84649
DGAT2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686A15125/HMFN1045
DGAT2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers