Learn More
Abnova™ Human DDX3X Partial ORF (NP_076829.1, 1 a.a. - 55 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00001654-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which interacts specifically with hepatitis C virus core protein resulting a change in intracellular location. This gene has a homolog located in the nonrecombining region of the Y chromosome. The protein sequence is 91% identical between this gene and the Y-linked homolog. [provided by RefSeq]
Sequence: MSHVAVENALGLDQQFAGLDLNSSDNQSGGSTASKGRYIPPHLRNREATRGFYDKSpecifications
NP_076829.1 | |
Liquid | |
1654 | |
DDX3X (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DBX/DDX14/DDX3/HLP2 | |
DDX3X | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
31.79kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSHVAVENALGLDQQFAGLDLNSSDNQSGGSTASKGRYIPPHLRNREATRGFYDK | |
RUO | |
DDX3X | |
Wheat Germ (in vitro) | |
GST |