missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DDX20 Full-length ORF (AAH34953.1, 1 a.a. - 824 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00011218-P02.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16136392

  • 508.00€ / 25µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which has an ATPase activity and is a component of the survival of motor neurons (SMN) complex. This protein interacts directly with SMN, the spinal muscular atrophy gene product, and may play a catalytic role in the function of the SMN complex on RNPs. [provided by RefSeq]

Sequence: MAAAFEASGALAAVATAMPAEHVAVQVPAPEPTPGPVRILRTAQDLSSPRTRTGDVLLAEPADFESLLLSRPVLEGLRAAGFERPSPVQLKAIPLGRCGLDLIVQAKSGTGKTCVFSTIALDSLVLENLSTQILILAPTREIAVQIHSVITAIGIKMEGLECHVFIGGTPLSQDKTRLKKCHIAVGSPGRIKQLIELDYLNPGSIRLFILDEADKLLEEGSFQEQINWIYSSLPASKQMLAVSATYPEFLANALTKYMRDPTFVRLNSSDPSLIGLKQYYKVVNSYPLAHKVFEEKTQHLQELFSRIPFNQALVFSNLHSRAQHLADILSSKGFPAECISGNMNQNQRLDAMAKLKHFHCRVLISTDLTSRGIDAEKVNLVVNLDVPLDWETYMHRIGRAGRFGTLGLTVTYCCRGEEGNMMMRIAQKCNINLLPLPDPIPSGLMEECVDWDVEVKAAVHTYGIASVPNQPLKKQIQKIERTLQIQKAHGDHMASSRNNSVSGLSVKSKNNTKQKLPVKSHSECGIIEKATSPKELGCDRQSEEQMKNSVQTPVENSTNSQHQVKEALPVSLPQIPCLSSFKIHQPYTLTFAELVEDYEHYIKEGLEKPVEIIRHYTGPGDQTVNPQNGFVRNKVTEQRVPVLASSSQSGDSESDSDSYSSRTSSQSKGNKSYLEGSSDNQLKDSESTPVDDRISLEQPPNGSDTPNPEKYQESPGIQMKTRLKEGASQRAKQSRRNLPRRSSFRLQTEAQEDDWYDCHREIRLSFSDTYQDYEEYWRAYYRAWQEYYAAASHSYYWNAQRHPSWMAAYHMNTIYLQEMMHSNQ
Specifications

Specifications

AAH34953.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
118.6kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp434H052/DP103/GEMIN3
DDX20
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
11218
DDX20 (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAAAFEASGALAAVATAMPAEHVAVQVPAPEPTPGPVRILRTAQDLSSPRTRTGDVLLAEPADFESLLLSRPVLEGLRAAGFERPSPVQLKAIPLGRCGLDLIVQAKSGTGKTCVFSTIALDSLVLENLSTQILILAPTREIAVQIHSVITAIGIKMEGLECHVFIGGTPLSQDKTRLKKCHIAVGSPGRIKQLIELDYLNPGSIRLFILDEADKLLEEGSFQEQINWIYSSLPASKQMLAVSATYPEFLANALTKYMRDPTFVRLNSSDPSLIGLKQYYKVVNSYPLAHKVFEEKTQHLQELFSRIPFNQALVFSNLHSRAQHLADILSSKGFPAECISGNMNQNQRLDAMAKLKHFHCRVLISTDLTSRGIDAEKVNLVVNLDVPLDWETYMHRIGRAGRFGTLGLTVTYCCRGEEGNMMMRIAQKCNINLLPLPDPIPSGLMEECVDWDVEVKAAVHTYGIASVPNQPLKKQIQKIERTLQIQKAHGDHMASSRNNSVSGLSVKSKNNTKQKLPVKSHSECGIIEKATSPKELGCDRQSEEQMKNSVQTPVENSTNSQHQVKEALPVSLPQIPCLSSFKIHQPYTLTFAELVEDYEHYIKEGLEKPVEIIRHYTGPGDQTVNPQNGFVRNKVTEQRVPVLASSSQSGDSESDSDSYSSRTSSQSKGNKSYLEGSSDNQLKDSESTPVDDRISLEQPPNGSDTPNPEKYQESPGIQMKTRLKEGASQRAKQSRRNLPRRSSFRLQTEAQEDDWYDCHREIRLSFSDTYQDYEEYWRAYYRAWQEYYAAASHSYYWNAQRHPSWMAAYHMNTIYLQEMMHSNQ
RUO
DDX20
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers