missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human DDIT3 Partial ORF (AAH03637.1, 94 a.a. - 155 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00001649-Q02.25ug
Additional Details : Weight : 0.02000kg
Description
Sequence: EEEEDQGRTRKRKQSGHSPARAGKQRMKEKEQENERKVAQLAEENERLKQEIERLTREVEATSpecifications
AAH03637.1 | |
Liquid | |
1649 | |
DDIT3 (Human) Recombinant Protein (Q02) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CEBPZ/CHOP/CHOP10/GADD153/MGC4154 | |
DDIT3 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.45kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EEEEDQGRTRKRKQSGHSPARAGKQRMKEKEQENERKVAQLAEENERLKQEIERLTREVEAT | |
RUO | |
DDIT3 | |
Wheat Germ (in vitro) | |
GST |