Learn More
Abnova™ Human DCLRE1C Partial ORF (NP_071932.2, 481 a.a. - 577 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00064421-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a nuclear protein that is involved in V(D)J recombination and DNA repair. The protein has single-strand-specific 5'-3' exonuclease activity; it also exhibits endonuclease activity on 5' and 3' overhangs and hairpins when complexed with protein kinase, DNA-activated, catalytic polypeptide. Mutations in this gene cause Athabascan-type severe combined immunodeficiency (SCIDA). [provided by RefSeq]
Sequence: ICPKDTYSDLKSRDKDVTIVPSTGEPTTLSSETHIPEEKSLLNLSTNADSQSSSDFEVPSTPEAELPKREHLQYLYEKLATGESIAVKKRKCSLLDTSpecifications
NP_071932.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.41kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ICPKDTYSDLKSRDKDVTIVPSTGEPTTLSSETHIPEEKSLLNLSTNADSQSSSDFEVPSTPEAELPKREHLQYLYEKLATGESIAVKKRKCSLLDT | |
RUO | |
DCLRE1C | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
64421 | |
DCLRE1C (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
A-SCID/DCLREC1C/FLJ11360/FLJ36438/RS-SCID/SCIDA/SNM1C | |
DCLRE1C | |
Recombinant | |
wheat germ expression system |