Learn More
R&D Systems™ Human Cytokine A Premixed Magnetic Luminex™ Performance Assay
Product Code. p-200051111
Click to view available options
Quantity:
10 Plex
11 Plex
12 Plex
13 Plex
14 Plex
15 Plex
16 Plex
17 Plex
18 Plex
19 Plex
2 Plex
20 Plex
21 Plex
22 Plex
3 Plex
4 Plex
5 Plex
6 Plex
7 Plex
8 Plex
9 Plex
Unit Size:
1 set
Each
Product Code. 18422100
Brand: R&D Systems™ FCSTM0319
Magnetic bead-based multiplex assay for the Luminex™ platform
TRUSTED_SUSTAINABILITY
Specifications
Quantity 19 Plex
Species Human
For Use With (Application) ELISA
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.