Learn More
Abnova™ Human CYTH2 Full-length ORF (NP_059431.1, 1 a.a. - 400 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009266-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the PSCD family. Members of this family have identical structural organization that consists of an N-terminal coiled-coil motif, a central Sec7 domain, and a C-terminal pleckstrin homology (PH) domain. The coiled-coil motif is involved in homodimerization, the Sec7 domain contains guanine-nucleotide exchange protein (GEP) activity, and the PH domain interacts with phospholipids and is responsible for association of PSCDs with membranes. Members of this family appear to mediate the regulation of protein sorting and membrane trafficking. The encoded protein exhibits GEP activity in vitro with ARF1, ARF3, and ARF6 and is 83% homologous to CYTH1. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MEDGVYEPPDLTPEERMELENIRRRKQELLVEIQRLREELSEAMSEVEGLEANEGSKTLQRNRKMAMGRKKFNMDPKKGIQFLVENELLQNTPEEIARFLYKGEGLNKTAIGDYLGEREELNLAVLHAFVDLHEFTDLNLVQALRQFLWSFRLPGEAQKIDRMMEAFAQRYCLCNPGVFQSTDTCYVLSFAVIMLNTSLHNPNVRDKPGLERFVAMNRGINEGGDLPEELLRNLYDSIRNEPFKIPEDDGNDLTHTFFNPDREGWLLKLGGGRVKTWKRRWFILTDNCLYYFEYTTDKEPRGIIPLENLSIREVDDPRKPNCFELYIPNNKGQLIKACKTEADGRVVEGNHMVYRISAPTQEEKDEWIKSIQAAVSVDPFYEMLAARKKRISVKKKQEQPSpecifications
NP_059431.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
72.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARNO/CTS18/CTS18.1/PSCD2/PSCD2L/SEC7L/Sec7p-L/Sec7p-like | |
CYTH2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
9266 | |
CYTH2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEDGVYEPPDLTPEERMELENIRRRKQELLVEIQRLREELSEAMSEVEGLEANEGSKTLQRNRKMAMGRKKFNMDPKKGIQFLVENELLQNTPEEIARFLYKGEGLNKTAIGDYLGEREELNLAVLHAFVDLHEFTDLNLVQALRQFLWSFRLPGEAQKIDRMMEAFAQRYCLCNPGVFQSTDTCYVLSFAVIMLNTSLHNPNVRDKPGLERFVAMNRGINEGGDLPEELLRNLYDSIRNEPFKIPEDDGNDLTHTFFNPDREGWLLKLGGGRVKTWKRRWFILTDNCLYYFEYTTDKEPRGIIPLENLSIREVDDPRKPNCFELYIPNNKGQLIKACKTEADGRVVEGNHMVYRISAPTQEEKDEWIKSIQAAVSVDPFYEMLAARKKRISVKKKQEQP | |
RUO | |
CYTH2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |