Learn More
ACROBiosystems Human Cystatin S / CST4 Protein, His Tag
Brand: ACROBiosystems CS4-H5223-100UG
Additional Details : Weight : 0.00001kg
Description
Cystatin-4 (CST4) is also known as Cystatin-S, Salivary acidic protein 1, which is a secreted protein, which belongs to the cystatin family. Cystatin-S is expressed in submandibular and sublingual saliva but not in parotid saliva (at protein level) and is also expressed in saliva, tears, urine and seminal fluid. Cystatin-4 / Cystatin S strongly inhibits papain and ficin, partially inhibits stem bromelain and bovine cathepsin C, but does not inhibit porcine cathepsin B or clostripain. Papain is inhibited non-competitively.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.0 kDa. The protein migrates as 15 kDa under reducing (R) condition (SDS-PAGE).Specifications
AAH65714 | |
100 μg | |
Ser 21 - Ala 141 | |
CST4,Cystatin-4,Cystatin-S,Cystatin-S5,Cystatin-SA-III | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
15.0 kDa | |
Human Cystatin S, His Tag (CS4-H5223) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Ala 141 (Accession # AAH65714). | |
-20°C | |
Cystatin S | |
Human | |
25 mM MES, 150 mM NaCl, pH6.0 | |
HEK293 | |
Other Recombinant Proteins |