Learn More
Abnova™ Human CYP2U1 Full-length ORF (AAH12027.1, 1 a.a. - 168 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00113612-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This enzyme is a hydroxylase that metabolizes arachidonic acid, docosahexaenoic acid, and other long chain fatty acids. [provided by RefSeq]
Sequence: MSSPGPSQPPAEDPPWPARLLRAPLGLLRLDPSGGALLLCGLVALLGWSWLRRRRARGIPPGPTPWPLVGNFGHVLLPPFLRRRSWLSSRTRAAGIDPSVIGPQVLLAHLARVYGSIFSFFIGHYLVVVLSDFHSVREALVQQAEVFSDRPRVPLISIVTKEKELFQESpecifications
AAH12027.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
45kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
P450TEC | |
CYP2U1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
113612 | |
CYP2U1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSSPGPSQPPAEDPPWPARLLRAPLGLLRLDPSGGALLLCGLVALLGWSWLRRRRARGIPPGPTPWPLVGNFGHVLLPPFLRRRSWLSSRTRAAGIDPSVIGPQVLLAHLARVYGSIFSFFIGHYLVVVLSDFHSVREALVQQAEVFSDRPRVPLISIVTKEKELFQE | |
RUO | |
CYP2U1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |