missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CXCL5/ENA-78 DuoSet ELISA

Product Code. 30000936
Change view
Click to view available options
Quantity:
1 Kit (5 Plates)
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30000936 1 Kit (5 Plates) Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30000936 Supplier R&D Systems™ Supplier No. DY25405
  • ONLINE EXCLUSIVE PRICE
    376.00€
    354.90€ / Pack of 5
    Save 21.10€
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Human CXCL5/ENA-78 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human CXCL5/ENA-78 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human CXCL5/ENA-78.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 15.60 to 1000 pg/mL
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 6374
Gene Symbol CXCL5
Host Species Human
Quantity 1 Kit (5 Plates)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.