missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CX3CR1 Partial ORF (AAH28078, 1 a.a. - 36 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00001524-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16163781

  • 335.00€ / 10µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

Sequence: MDQFPESVTENFEYDDLAEACYIGDIVVFGTVFLSI
Specifications

Specifications

AAH28078
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
29.7kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MDQFPESVTENFEYDDLAEACYIGDIVVFGTVFLSI
RUO
CX3CR1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
1524
CX3CR1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CCRL1/CMKBRL1/CMKDR1/GPR13/GPRV28/V28
CX3CR1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers