missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CST8 Full-length ORF (AAH69536.1, 1 a.a. - 90 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010047-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16124412

  • 508.00€ / 25µg
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

The cystatin superfamily encompasses proteins that contain multiple cystatin-like sequences. Some of the members are active cysteine protease inhibitors, while others have lost or perhaps never acquired this inhibitory activity. There are three inhibitory families in the superfamily, including the type 1 cystatins (stefins), type 2 cystatins and the kininogens. The type 2 cystatin proteins are a class of cysteine proteinase inhibitors found in a variety of human fluids and secretions. The cystatin locus on chromosome 20 contains the majority of the type 2 cystatin genes and pseudogenes. This gene is located in the cystatin locus and encodes a protein similar to type 2 cystatins. The protein exhibits highly tissue-specific expression in the reproductive tract, suggesting implicit roles in reproduction. Alternative splicing identified in mouse is suggested in human based on EST evidence but the full-length nature of putative variants has not been determined. [provided by RefSeq]

Sequence: MQEYNKESEDKYVFLVVKTLQAQLQVTNLLEYLIDVEIARSDCRKPLSTNEICAIQENSKLKRKLSCSFLVGALPWNGEFTVMEKKCEDA
Specifications

Specifications

AAH69536.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.8kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CRES
CST8
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
10047
CST8 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MQEYNKESEDKYVFLVVKTLQAQLQVTNLLEYLIDVEIARSDCRKPLSTNEICAIQENSKLKRKLSCSFLVGALPWNGEFTVMEKKCEDA
RUO
CST8
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers