missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CSPG3 Partial ORF (NP_004377, 81 a.a. - 179 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00001463-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Neurocan is a chondroitin sulfate proteoglycan thought to be involved in the modulation of cell adhesion and migration.[supplied by OMIM]
Sequence: VRTASGQRQDLPILVAKDNVVRVAKSWQGRVSLPSYPRRRANATLLLGPLRASDSGLYRCQVVRGIEDEQDLVPLEVTGVVFHYRSARDRYALTFAEAQSpecifications
NP_004377 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VRTASGQRQDLPILVAKDNVVRVAKSWQGRVSLPSYPRRRANATLLLGPLRASDSGLYRCQVVRGIEDEQDLVPLEVTGVVFHYRSARDRYALTFAEAQ | |
RUO | |
NCAN | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
1463 | |
CSPG3 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CSPG3/FLJ44681 | |
NCAN | |
Recombinant | |
wheat germ expression system |