missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CRTAP Partial ORF (NP_006362, 307 a.a. - 401 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010491-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192636

  • 335.00€ / 10µg
Estimated Shipment: 24-09-2024
to see stock.
Add to basket

Description

Description

The protein encoded by this gene is similar to the chicken and mouse CRTAP genes. The encoded protein is a scaffolding protein that may influence the activity of at least one member of the cytohesin/ARNO family in response to specific cellular stimuli. Defects in this gene are associated with osteogenesis imperfecta, a connective tissue disorder characterized by bone fragility and low bone mass. [provided by RefSeq]

Sequence: KLNDLKNAAPCAVSYLLFDQNDKVMQQNLVYYQYHRDTWGLSDEHFQPRPEAVQFFNVTTLQKELYDFAKENIMDDDEGEVVEYVDDLLELEETS
Specifications

Specifications

NP_006362
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.19kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KLNDLKNAAPCAVSYLLFDQNDKVMQQNLVYYQYHRDTWGLSDEHFQPRPEAVQFFNVTTLQKELYDFAKENIMDDDEGEVVEYVDDLLELEETS
RUO
CRTAP
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
10491
CRTAP (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CASP/LEPREL3
CRTAP
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers