Learn More
Abnova™ Human CRH Partial ORF (AAH11031, 154 a.a. - 196 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00001392-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Corticotropin-releasing hormone (CRH) is a 41-amino acid peptide derived from a 191-amino acid preprohormone. CRH is secreted by the paraventricular nucleus (PVN) of the hypothalamus in response to stress. Marked reduction in CRH has been observed in association with Alzheimer disease and autosomal recessive hypothalamic corticotropin dificiency has multiple and potentially fatal metabolic consequences including hypoglycemia and hepatitis. In addition to production in the hypothalamus, CRH is also synthesized in peripheral tissues, such as T lymphocytes and is highly expressed in the placenta. In the placenta CRH is a marker that determines the length of gestation and the timing of parturition and delivery. A rapid increase in circulating levels of CRH occurs at the onset of parturition, suggesting that, in addition to its metabolic functions, CRH may act as a trigger for parturition. [provided by RefSeq]
Sequence: SEEPPISLDLTFHLLREVLEMARAEQLAQQAHSNRKLMEIIGKSpecifications
AAH11031 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
30.47kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SEEPPISLDLTFHLLREVLEMARAEQLAQQAHSNRKLMEIIGK | |
RUO | |
CRH | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
1392 | |
CRH (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CRF | |
CRH | |
Recombinant | |
wheat germ expression system |