missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CPXM2 Partial ORF (NP_937791, 190 a.a. - 279 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00119587-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16121627

  • 545.00€ / 25µg

Estimated Shipment: 23-09-2022
to see stock.
Add to basket

Description

Description

Sequence: DLQQWIEVDARRLTRFTGVITQGRNSLWLSDWVTSYKVMVSNDSHTWVTVKNGSGDMIFEGNSEKEIPVLNELPVPMVARYIRINPQSWF
Specifications

Specifications

NP_937791
Liquid
119587
CPXM2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
UNQ676
CPXM2
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.64kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
DLQQWIEVDARRLTRFTGVITQGRNSLWLSDWVTSYKVMVSNDSHTWVTVKNGSGDMIFEGNSEKEIPVLNELPVPMVARYIRINPQSWF
RUO
CPXM2
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers