Learn More
Abnova™ Human CPM Full-length ORF (NP_001005502.1, 1 a.a. - 443 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00001368-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a membrane-bound arginine/lysine carboxypeptidase. Its expression is associated with monocyte to macrophage differentiation. This encoded protein contains hydrophobic regions at the amino and carboxy termini and has 6 potential asparagine-linked glycosylation sites. The active site residues of carboxypeptidases A and B are conserved in this protein. Three alternatively spliced transcript variants encoding the same protein have been described for this gene. [provided by RefSeq]
Sequence: MDFPCLWLGLLLPLVAALDFNYHRQEGMEAFLKTVAQNYSSVTHLHSIGKSVKGRNLWVLVVGRFPKEHRIGIPEFKYVANMHGDETVGRELLLHLIDYLVTSDGKDPEITNLINSTRIHIMPSMNPDGFEAVKKPDCYYSIGRENYNQYDLNRNFPDAFEYNNVSRQPETVAVMKWLKTETFVLSANLHGGALVASYPFDNGVQATGALYSRSLTPDDDVFQYLAHTYASRNPNMKKGDECKNKMNFPNGVTNGYSWYPLQGGMQDYNYIWAQCFEITLELSCCKYPREEKLPSFWNNNKASLIEYIKQVHLGVKGQVFDQNGNPLPNVIVEVQDRKHICPYRTNKYGEYYLLLLPGSYIINVTVPGHDPHITKVIIPEKSQNFSALKKDILLPFQGQLDSIPVSNPSCPMIPLYRNLPDHSAATKPSLFLFLVSLLHIFFKSpecifications
NP_001005502.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
76.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CPM | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
1368 | |
CPM (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDFPCLWLGLLLPLVAALDFNYHRQEGMEAFLKTVAQNYSSVTHLHSIGKSVKGRNLWVLVVGRFPKEHRIGIPEFKYVANMHGDETVGRELLLHLIDYLVTSDGKDPEITNLINSTRIHIMPSMNPDGFEAVKKPDCYYSIGRENYNQYDLNRNFPDAFEYNNVSRQPETVAVMKWLKTETFVLSANLHGGALVASYPFDNGVQATGALYSRSLTPDDDVFQYLAHTYASRNPNMKKGDECKNKMNFPNGVTNGYSWYPLQGGMQDYNYIWAQCFEITLELSCCKYPREEKLPSFWNNNKASLIEYIKQVHLGVKGQVFDQNGNPLPNVIVEVQDRKHICPYRTNKYGEYYLLLLPGSYIINVTVPGHDPHITKVIIPEKSQNFSALKKDILLPFQGQLDSIPVSNPSCPMIPLYRNLPDHSAATKPSLFLFLVSLLHIFFK | |
RUO | |
CPM | |
Recombinant | |
wheat germ expression system |