missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human COX8C Full-length ORF (NP_892016.1, 1 a.a. - 72 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00341947-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16160214

  • 335.00€ / 10µg
Estimated Shipment: 18-06-2024
to see stock.
Add to basket

Description

Description

Sequence: MPLLRGRCPARRHYRRLALLGLQPAPRFAHSGPPRQRPLSAAEMAVGLVVFFTTFLTPAAYVLGNLKQFRRN
Specifications

Specifications

NP_892016.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
COX8-3/MGC119774/MGC119775
COX8C
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
341947
COX8C (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MPLLRGRCPARRHYRRLALLGLQPAPRFAHSGPPRQRPLSAAEMAVGLVVFFTTFLTPAAYVLGNLKQFRRN
RUO
COX8C
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers