missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CORO1A Partial ORF (NP_009005, 360 a.a. - 461 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00011151-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: QEDLYPPTAGPDPALTAEEWLGGRDAGPLLISLKDGYVPPKSRELRVNRGLDTGRRRAAPEASGTPSSDAVSRLEEEMRKLQATVQELQKRLDRLEETVQAKSpecifications
NP_009005 | |
Liquid | |
11151 | |
CORO1A (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CLABP/CLIPINA/FLJ41407/HCORO1/MGC117380/TACO/p57 | |
CORO1A | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.96kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QEDLYPPTAGPDPALTAEEWLGGRDAGPLLISLKDGYVPPKSRELRVNRGLDTGRRRAAPEASGTPSSDAVSRLEEEMRKLQATVQELQKRLDRLEETVQAK | |
RUO | |
CORO1A | |
Wheat Germ (in vitro) | |
GST |