Learn More
Abnova™ Human COPZ2 Full-length ORF (NP_057513.1, 1 a.a. - 210 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00051226-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a subunit of the coatomer protein complex, a seven-subunit complex that functions in the formation of COPI-type, non-clathrin-coated vesicles. COPI vesicles function in the retrograde Golgi-to-ER transport of dilysine-tagged proteins. This gene is similar to a related family member, and the two encoded proteins form distinct isotypes of the coatomer protein complex. [provided by RefSeq]
Sequence: MQRPEAWPRPHPGEGAAAAQAGGPAPPARAGEPSGLRLQEPSLYTIKAVFILDNDGRRLLAKYYDDTFPSMKEQMVFEKNVFNKTSRTESEIAFFGGMTIVYKNSIDLFLYVVGSSYENELMLMSVLTCLFESLNHMLRKNVEKRWLLENMDGAFLVLDEIVDGGVILESDPQQVIQKVNFRADDGGLTEQSVAQVLQSAKEQIKWSLLKSpecifications
NP_057513.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
49.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC23008 | |
COPZ2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
51226 | |
COPZ2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MQRPEAWPRPHPGEGAAAAQAGGPAPPARAGEPSGLRLQEPSLYTIKAVFILDNDGRRLLAKYYDDTFPSMKEQMVFEKNVFNKTSRTESEIAFFGGMTIVYKNSIDLFLYVVGSSYENELMLMSVLTCLFESLNHMLRKNVEKRWLLENMDGAFLVLDEIVDGGVILESDPQQVIQKVNFRADDGGLTEQSVAQVLQSAKEQIKWSLLK | |
RUO | |
COPZ2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |