missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human COL11A2 Partial ORF (NP_542411, 29 a.a. - 128 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00001302-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16193391

  • 528.00€ / 25µg

Estimated Shipment: 07-11-2023
to see stock.
Add to basket

Description

Description

This gene encodes one of the two alpha chains of type XI collagen, a minor fibrillar collagen. It is located on chromosome 6 very close to but separate from the gene for retinoid X receptor beta. Type XI collagen is a heterotrimer but the third alpha chain is a post-translationally modified alpha 1 type II chain. Proteolytic processing of this type XI chain produces PARP, a proline/arginine-rich protein that is an amino terminal domain. Mutations in this gene are associated with type III Stickler syndrome, otospondylomegaepiphyseal dysplasia (OSMED syndrome), Weissenbacher-Zweymuller syndrome, autosomal dominant non-syndromic sensorineural type 13 deafness (DFNA13), and autosomal recessive non-syndromic sensorineural type 53 deafness (DFNB53). Alternative splicing results in multiple transcript variants. A related pseudogene is located nearby on chromosome 6. [provided by RefSeq]

Sequence: APPVDVLRALRFPSLPDGVRRAKGICPADVAYRVARPAQLSAPTRQLFPGGFPKDFSLLTVVRTRPGLQAPLLTLYSAQGVRQLGLELGRPVRFLYEDQT
Specifications

Specifications

NP_542411
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
APPVDVLRALRFPSLPDGVRRAKGICPADVAYRVARPAQLSAPTRQLFPGGFPKDFSLLTVVRTRPGLQAPLLTLYSAQGVRQLGLELGRPVRFLYEDQT
RUO
COL11A2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
1302
COL11A2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DFNA13/DFNB53/HKE5/PARP/STL3
COL11A2
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers