Learn More
Abnova™ Human COG8 Partial ORF (NP_115758.2, 450 a.a. - 545 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00084342-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein that is a component of the conserved oligomeric Golgi (COG) complex, a multiprotein complex that plays a structural role in the Golgi apparatus, and is involved in intracellular membrane trafficking and glycoprotein modification. Mutations in this gene cause congenital disorder of glycosylation, type IIh, a disease that is characterized by under-glycosylated serum proteins, and whose symptoms include severe psychomotor retardation, failure to thrive, seizures, and dairy and wheat product intolerance. [provided by RefSeq]
Sequence: RLCCPVALAQDVTGALEDALAKVTKIILAFHRAEEAAFSSGEQELFVQFCTVFLEDLVPYLNRCLQVLFPPAQIAQTLGIPPTQLSKYGNLGHVNISpecifications
NP_115758.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.3kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RLCCPVALAQDVTGALEDALAKVTKIILAFHRAEEAAFSSGEQELFVQFCTVFLEDLVPYLNRCLQVLFPPAQIAQTLGIPPTQLSKYGNLGHVNI | |
RUO | |
COG8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
84342 | |
COG8 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DOR1/FLJ22315 | |
COG8 | |
Recombinant | |
wheat germ expression system |