Learn More
Abnova™ Human CNTNAP4 Partial ORF (NP_207837, 1145 a.a. - 1244 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00085445-Q01.25ug
Additional Details : Weight : 0.00010kg
Beschreibung
This gene product belongs to the neurexin family, members of which function in the vertebrate nervous system as cell adhesion molecules and receptors. This protein, like other neurexin proteins, contains epidermal growth factor repeats and laminin G domains. In addition, it includes an F5/8 type C domain, discoidin/neuropilin- and fibrinogen-like domains, and thrombospondin N-terminal-like domains. Alternative splicing results in two transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: VLGRILEHSDVDQETALAGAQGFTGCLSAVQLSHVAPLKAALHPSHPDPVTVTGHVTESSCMAQPGTDATSRERTHSFADHSGTIDDREPLANAIKSDSASpezifikation
NP_207837 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VLGRILEHSDVDQETALAGAQGFTGCLSAVQLSHVAPLKAALHPSHPDPVTVTGHVTESSCMAQPGTDATSRERTHSFADHSGTIDDREPLANAIKSDSA | |
RUO | |
CNTNAP4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
85445 | |
CNTNAP4 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CASPR4/KIAA1763 | |
CNTNAP4 | |
Recombinant | |
wheat germ expression system |