missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CNTN4 Partial ORF (NP_783200.1, 800 a.a. - 898 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00152330-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192167

  • 349.00€ / 10µg

Estimated Shipment: 10-11-2023
to see stock.
Add to basket

Description

Description

The protein encoded by this gene is a member of the immunoglobulin superfamily. It is a glycosylphosphatidylinositol (GPI)-anchored neuronal membrane protein that functions as a cell adhesion molecule. It may play a role in the formation of axon connections in the developing nervous system. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined. [provided by RefSeq]

Sequence: EPTKPPASIFARSLSATDIEVFWASPLEKNRGRIQGYEVKYWRHEDKEENARKIRTVGNQTSTKITNLKGSVLYHLAVKAYNSAGTGPSSATVNVTTRK
Specifications

Specifications

NP_783200.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
EPTKPPASIFARSLSATDIEVFWASPLEKNRGRIQGYEVKYWRHEDKEENARKIRTVGNQTSTKITNLKGSVLYHLAVKAYNSAGTGPSSATVNVTTRK
RUO
CNTN4
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
152330
CNTN4 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AXCAM/BIG-2/CNTN4A/MGC33615
CNTN4
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers