Learn More
ACROBiosystems Human CNTF R alpha Protein, His Tag
Brand: ACROBiosystems CNR-H5225-100UG
Additional Details : Weight : 0.00001kg
Description
Ciliary neurotrophic factor receptor subunit alpha (CNTFR or CNTFR-alpha) belongs to the type I cytokine receptor family and type 3 subfamily. CNTFR contains two fibronectin type-III domains and one Ig-like C2-type (immunoglobulin-like) domain. This receptor and its cognate ligand support the survival of neurons. This receptor is most closely related to the interleukin-6 receptor. This receptor possesses an unusual attachment to the cell membrane through a glycophosphatidylinositol linkage.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.6 kDa. The protein migrates as 45-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
AAH01492 | |
100 μg | |
Gln 23 - Pro 346 | |
CNTFR,CNTF R alpha | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
37.6 kDa | |
Human CNTF R alpha, His Tag (CNR-H5225) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Pro 346 (Accession # AAH01492). | |
-20°C | |
CNTF R alpha | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |