Learn More
Abnova™ Human CNOT6 Partial ORF (NP_056270.2, 112 a.a. - 210 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00057472-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a subunit of the CCR4-NOT core transcriptional regulation complex. The encoded protein has a 3'-5' RNase activity and prefers polyadenylated substates. [provided by RefSeq]
Sequence: LPFELGKLFQLQTLGLKGNPLTQDILNLYQEPDGTRRLLNYLLDNLSGTAKRITTEQPPPRSWIMLQEPDRTRPTALFSVMCYNVLCDKYATRQLYGYCSpecifications
NP_056270.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LPFELGKLFQLQTLGLKGNPLTQDILNLYQEPDGTRRLLNYLLDNLSGTAKRITTEQPPPRSWIMLQEPDRTRPTALFSVMCYNVLCDKYATRQLYGYC | |
RUO | |
CNOT6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
57472 | |
CNOT6 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CCR4/KIAA1194 | |
CNOT6 | |
Recombinant | |
wheat germ expression system |