missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CNN2 Partial ORF (NP_004359.1, 193 a.a. - 290 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00001265-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16156484

  • 335.00€ / 10µg
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

The protein encoded by this gene, which can bind actin, calmodulin, troponin C, and tropomyosin, may function in the structural organization of actin filaments. The encoded protein could play a role in smooth muscle contraction and cell adhesion. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: PKNHILPPMDHSTISLQMGTNKCASQVGMTAPGTRRHIYDTKLGTDKCDNSSMSLQMGYTQGANQSGQVFGLGRQIYDPKYCPQGTVADGAPSGTGDC
Specifications

Specifications

NP_004359.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PKNHILPPMDHSTISLQMGTNKCASQVGMTAPGTRRHIYDTKLGTDKCDNSSMSLQMGYTQGANQSGQVFGLGRQIYDPKYCPQGTVADGAPSGTGDC
RUO
CNN2
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
1265
CNN2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CNN2
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers