Learn More
Abnova™ Human CML2 Partial ORF (NP_057431.1, 83 a.a. - 179 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00051471-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is highly similar to the N-acetyltransferase 8 (NAT8) gene product, which is a kidney and liver protein with homology to bacterial acetyltransferases involved in drug resistance. This gene is localized on chromosome 2 in the vicinity of the NAT8 gene and may represent a pseudogene of NAT8. This gene contains two polymorphic nonsense mutations that disrupt the active site of the protein. The full-length product of this gene contains a complete acetyltransferase domain and is identical in length to NAT8. [provided by RefSeq]
Sequence: TRYVDIALRTDMSDITKSYLSECGSCFWVGESEEKVVGTVGALPVDDPTLREKRLQLFHLSVDNEHRGQGIAKALVRTVLQFARDQGYSEVVLDTSNSpecifications
NP_057431.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.41kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
TRYVDIALRTDMSDITKSYLSECGSCFWVGESEEKVVGTVGALPVDDPTLREKRLQLFHLSVDNEHRGQGIAKALVRTVLQFARDQGYSEVVLDTSN | |
RUO | |
NAT8B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
51471 | |
CML2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CML2/Hcml2/MGC97061/NAT8BP | |
NAT8B | |
Recombinant | |
wheat germ expression system |