missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CLSTN1 Partial ORF (NP_001009566.1, 759 a.a. - 858 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00022883-Q02.25ug
Additional Details : Weight : 0.02000kg
Description
Sequence: FTGVDTMASYEEVLHLLRYRNWHARSLLDRKFKLICSELNGRYISNEFKVEVNVIHTANPMEHANHMAAQPQFVHPEHRSFVDLSGHNLANPHPFAVVPSSpecifications
NP_001009566.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
FTGVDTMASYEEVLHLLRYRNWHARSLLDRKFKLICSELNGRYISNEFKVEVNVIHTANPMEHANHMAAQPQFVHPEHRSFVDLSGHNLANPHPFAVVPS | |
RUO | |
CLSTN1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
22883 | |
CLSTN1 (Human) Recombinant Protein (Q02) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ALC-ALPHA/CSTN1/FLJ32258/KIAA0911/PIK3CD/XB31alpha/alcalpha1/alcalpha2 | |
CLSTN1 | |
Recombinant | |
wheat germ expression system |