missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CLIP2 Partial ORF (NP_003379, 946 a.a. - 1046 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007461-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16157515

  • 508.00€ / 25µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

The protein encoded by this gene belongs to the family of cytoplasmic linker proteins, which have been proposed to mediate the interaction between specific membranous organelles and microtubules. This protein was found to associate with both microtubules and an organelle called the dendritic lamellar body. This gene is hemizygously deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.23. Alternative splicing of this gene generates 2 transcript variants. [provided by RefSeq]

Sequence: LKDDIRGLREKLTGLDKEKSLSDQRRYSLIDRSSAPELLRLQHQLMSTEDALRDALDQAQQVEKLMEAMRSCPDKAQTIGNSGSANGIHQQDKAQKQEDKH
Specifications

Specifications

NP_003379
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.85kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LKDDIRGLREKLTGLDKEKSLSDQRRYSLIDRSSAPELLRLQHQLMSTEDALRDALDQAQQVEKLMEAMRSCPDKAQTIGNSGSANGIHQQDKAQKQEDKH
RUO
CLIP2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
7461
CLIP2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CLIP/CLIP-115/CYLN2/KIAA0291/MGC11333/WBSCR3/WBSCR4/WSCR3/WSCR4
CLIP2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers