missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CLEC10A Partial ORF (NP_006335.2, 70 a.a. - 169 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010462-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192576

  • 528.00€ / 25µg

Estimated Shipment: 10-11-2023
to see stock.
Add to basket

Description

Description

This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type 2 transmembrane protein may function as a cell surface antigen. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]

Sequence: VTLRTDFSNFTSNTVAEIQALTSQGSSLEETIASLKAEVEGFKQERQAVHSEMLLRVQQLVQDLKKLTCQVATLNNNGEEASTEGTCCPVNWVEHQDSCY
Specifications

Specifications

NP_006335.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VTLRTDFSNFTSNTVAEIQALTSQGSSLEETIASLKAEVEGFKQERQAVHSEMLLRVQQLVQDLKKLTCQVATLNNNGEEASTEGTCCPVNWVEHQDSCY
RUO
CLEC10A
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
10462
CLEC10A (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CD301/CLECSF13/CLECSF14/HML/HML2
CLEC10A
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers