missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CLDN2 Partial ORF (NP_065117.1, 138 a.a. - 163 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009075-Q02.10ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16095115

  • 335.00€ / 10µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

Members of the claudin protein family, such as CLDN2, are expressed in an organ-specific manner and regulate the tissue-specific physiologic properties of tight junctions (Sakaguchi et al., 2002 [PubMed 11934881]).[supplied by OMIM]

Sequence: WNLHGILRDFYSPLVPDSMKFEIGEA
Specifications

Specifications

NP_065117.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
28.49kDa
12.5% SDS-PAGE Stained with Coomassie Blue
WNLHGILRDFYSPLVPDSMKFEIGEA
RUO
CLDN2
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
9075
CLDN2 (Human) Recombinant Protein (Q02)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CLDN2
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers