Learn More
Abnova™ Human CISH Full-length ORF (NP_659508.1, 1 a.a. - 258 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00001154-P02.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene contains a SH2 domain and a SOCS box domain. The protein thus belongs to the cytokine-induced STAT inhibitor (CIS), also known as suppressor of cytokine signaling (SOCS) or STAT-induced STAT inhibitor (SSI), protein family. CIS family members are known to be cytokine-inducible negative regulators of cytokine signaling. The expression of this gene can be induced by IL2, IL3, GM-CSF and EPO in hematopoietic cells. Proteasome-mediated degradation of this protein has been shown to be involved in the inactivation of the erythropoietin receptor. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MVLCVQGPRPLLAVERTGQRPLWAPSLELPKPVMQPLPAGAFLEEVAEGTPAQTESEPKVLDPEEDLLCIAKTFSYLRESGWYWGSITASEARQHLQKMPEGTFLVRDSTHPSYLFTLSVKTTRGPTNVRIEYADSSFRLDSNCLSRPRILAFPDVVSLVQHYVASCTADTRSDSPDPAPTPALPMPKEDAPSDPALPAPPPATAVHLKLVQPFVRRSSARSLQHLCRLVINRLVADVDCLPLPRRMADYLRQYPFQLSpecifications
NP_659508.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
55.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CIS/CIS-1/G18/SOCS | |
CISH | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
1154 | |
CISH (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MVLCVQGPRPLLAVERTGQRPLWAPSLELPKPVMQPLPAGAFLEEVAEGTPAQTESEPKVLDPEEDLLCIAKTFSYLRESGWYWGSITASEARQHLQKMPEGTFLVRDSTHPSYLFTLSVKTTRGPTNVRIEYADSSFRLDSNCLSRPRILAFPDVVSLVQHYVASCTADTRSDSPDPAPTPALPMPKEDAPSDPALPAPPPATAVHLKLVQPFVRRSSARSLQHLCRLVINRLVADVDCLPLPRRMADYLRQYPFQL | |
RUO | |
CISH | |
Wheat Germ (in vitro) | |
GST | |
Liquid |