missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CIR Partial ORF (NP_004873, 136 a.a. - 204 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009541-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16100906

  • 508.00€ / 25µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

Sequence: HVNTDRECPLFGLSGINASSVPTDGSGPSMHPSELIAEMRNSGFALKRNVLGRNLTANDPSQEYVASEG
Specifications

Specifications

NP_004873
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.33kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
HVNTDRECPLFGLSGINASSVPTDGSGPSMHPSELIAEMRNSGFALKRNVLGRNLTANDPSQEYVASEG
RUO
CIR
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
9541
CIR (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CIR
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers