missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CILP Partial ORF (NP_003604, 129 a.a. - 226 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008483-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16158625

  • 335.00€ / 10µg
Estimated Shipment: 14-06-2024
to see stock.
Add to basket

Description

Description

Major alterations in the composition of the cartilage extracellular matrix occur in joint disease, such as osteoarthrosis. This gene encodes the cartilage intermediate layer protein (CILP), which increases in early osteoarthrosis cartilage. The encoded protein was thought to encode a protein precursor for 2 different proteins, namely CILP and a homolog of NTPPHase, however later studies identified no nucleotide pyrophosphatase phosphodiesterase (NPP) activity. One isoform of the protein, CILP-1, functions as an IGF-1 antagonist. [provided by RefSeq]

Sequence: NCSNYTVRFLCPPGSLRRDTERIWSPWSPWSKCSAACGQTGVQTRTRICLAEMVSLCSEASEEGQHCMGQDCTACDLTCPMGQVNADCDACMCQDFML
Specifications

Specifications

NP_003604
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
NCSNYTVRFLCPPGSLRRDTERIWSPWSPWSKCSAACGQTGVQTRTRICLAEMVSLCSEASEEGQHCMGQDCTACDLTCPMGQVNADCDACMCQDFML
RUO
CILP
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
8483
CILP (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CILP-1/HsT18872
CILP
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers