missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CIDEB Partial ORF (NP_055245, 3 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00027141-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16125746

  • 359.00€ / 10µg

Estimated Shipment: 21-09-2022
to see stock.
Add to basket

Description

Description

Sequence: YLSALNPSDLLRSVSNISSEFGRRVWTSAPPPQRPFRVCDHKRTIRKGLTAATRQELLAKALETLLLNGVLTLVLEEDGTAVDSEDFFQLLEDDTCLMVLQSGQSWSP
Specifications

Specifications

NP_055245
Liquid
27141
CIDEB (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CIDEB
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.62kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
YLSALNPSDLLRSVSNISSEFGRRVWTSAPPPQRPFRVCDHKRTIRKGLTAATRQELLAKALETLLLNGVLTLVLEEDGTAVDSEDFFQLLEDDTCLMVLQSGQSWSP
RUO
CIDEB
Yes
wheat germ expression system
Videos
SDS
Documents
Certificates
Special Offers

Special Offers